Dipotassium tetraborate tetrahydrate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

93-1950-500G 93-1950-2000G
100205-390EA 77.85 USD
100205-390 100210-224
Dipotassium tetraborate tetrahydrate ≥99%
Dipotassium tetraborate tetrahydrate
Formula: B₄K₂O₇·4H₂O
Melting Pt: Dec.
Density: 1,74
MDL Number: MFCD00149924
CAS Number: 12045-78-2
EINECS: 215-575-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR