Potassium tellurite hydrate ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Potassium tellurite(IV) hydrate, Potassium tellurium trioxide hydrate, di-Potassium tellurium trioxide hydrate, Potassium tellurium (IV) oxide hydrate, Potassium tellurite monohydrate
93-1949-25G
93-1949-100G
93-1949-5G
100199-502EA
266.09
USD
100199-502
100205-864
100205-388
Potassium tellurite hydrate ≥97%
Potassium tellurite hydrate
Formula:
K₂O₃Te·H₂O MW: 271.81 g/mol Melting Pt: 465 °C (dec.) |
MDL Number:
MFCD00149923 CAS Number: 123333-66-4 EINECS: 232-213-1 UN: 3284 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...