≥98% ACS
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate
93-1940-250G
93-1940-1000G
100208-514EA
82.09
USD
100208-514
100201-076
≥98% ACS
di-Potassium hydrogen phosphate
Formula:
K₂HPO₄ MW: 174.18 g/mol Melting Pt: 340 Density: 2.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011383 CAS Number: 7758-11-4 EINECS: 231-834-5 Merck Index: 13,07743 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...