Potassium dihydrogen phosphate ≥99% ACS
Supplier: STREM CHEMICALS INC MS
93-1939-250G
93-1939-1000G
93-1939-5000G
100197-724EA
60.86
USD
100197-724
100209-034
100209-036
Potassium dihydrogen phosphate ≥99% ACS
Potassium dihydrogen phosphate
Formula:
KH₂PO₄ Boiling Pt: ∼100 °C (1013 hPa) Melting Pt: 253 °C Density: 2.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011401 CAS Number: 7778-77-0 EINECS: 231-913-4 REACH: 01-2119490224-41 Merck Index: 13,07744 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...