Potassium hexacyanoferrate(II) trihydrate ≥98.5% ACS
Supplier: STREM CHEMICALS INC MS
|
|
Synonyms:
Potassiumhexacyanoferrate(II) trihydrate, Tetrapotassium hexacyanoferrate trihydrate, Potassium ferrocyanide trihydrate
93-1921-500G
93-1921-100G
100199-498EA
179.75
USD
100199-498
100205-382
Potassium hexacyanoferrate(II) trihydrate ≥98.5% ACS
Potassium hexacyanoferrate(II) trihydrate
Formula:
K₄Fe(CN)₆·3H₂O MW: 422.39 g/mol Melting Pt: 70 (dec.) Density: 1.85 Storage Temperature: Ambient |
MDL Number:
MFCD00167023 CAS Number: 14459-95-1 EINECS: 237-722-2 Merck Index: 12,07793 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...