Potassium hexacyanoferrate(III) ≥99% ACS

Supplier: STREM CHEMICALS INC MS

Synonyms: Potassium ferricyanide(III), Tripotassium hexacyanoferrate, Potassium ferricyanide

93-1920-1000G 93-1920-250G
100204-678EA 329.78 USD
100204-678 100204-680
Potassium hexacyanoferrate(III) ≥99% ACS
Potassium hexacyanoferrate(III)
Formula: K₃Fe(CN)₆
MW: 329.25 g/mol
Melting Pt: Dec.
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011392
CAS Number: 13746-66-2
EINECS: 237-323-3
Merck Index: 13,07712

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR