Potassium tetrahydroborate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Potassium tetrahydridoborate, Potassium borohydride

93-1907-250G 93-1907-50G
100209-028EA 392.06 USD
100209-028 100200-524
Potassium tetrahydroborate ≥98%
Potassium tetrahydroborate
Formula: H₄BK
MW: 53.94 g/mol
Melting Pt: 500 (dec.)
Density: 1.178
Storage Temperature: Ambient
MDL Number: MFCD00011396
CAS Number: 13762-51-1
EINECS: 237-360-5
UN: 1870
ADR: 4.3 I
Merck Index: 12,07777

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR