Potassium hydrogen difluoride ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-1905-500G
93-1905-2000G
100204-198EA
83.51
USD
100204-198
100207-274
Potassium hydrogen difluoride ≥99%
Potassium hydrogen difluoride
Formula:
HF₂K Melting Pt: 239 °C Density: 2.37 g/cm³ (20 °C) |
MDL Number:
MFCD00011403 CAS Number: 7789-29-9 EINECS: 232-156-2 UN: 1811 ADR: 8,II Merck Index: 12,07771 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...