Potassium acetate ≥99% ACS

Supplier: STREM CHEMICALS INC MS

Synonyms: Acetic acid potassium salt

93-1901-2000G 93-1901-500G
100210-220EA 570.4 USD
100210-220 100205-376
Potassium acetate ≥99% ACS
Potassium acetate
Formula: CH₃COOK
MW: 98.14 g/mol
Melting Pt: 304
Density: 1.57 g/cm³ (25 °C)
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
EINECS: 204-822-2
Merck Index: 13,07687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR