Sulfur, powder, purified, precipitated
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-1617-250G
93-1617-1000G
100205-374EA
60.86
USD
100205-374
100210-218
Sulfur, powder, purified, precipitated
Sulphur
Formula:
S Boiling Pt: 444.67 °C Melting Pt: 112.8 °C Density: 2.36 g/cm³ (20 °C) Flash Pt: 405 °F Storage Temperature: Ambient |
MDL Number:
MFCD00085316 CAS Number: 7704-34-9 EINECS: 231-722-6 UN: 1350 ADR: 4.1,III Merck Index: 13,09059 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...