Phosphorus(III) chloride ≥99.998% (P basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Phosphorus trichloride

93-1588-10G 93-1588-50G
100200-728EA 133.05 USD
100200-728 100200-730
Phosphorus(III) chloride ≥99.998% (P basis), PURATREM
Phosphorus(III) chloride

Halides for CVD & ALD

Formula: Cl₃P
MW: 137.33 g/mol
Boiling Pt: 76
Melting Pt: -112
Density: 1.574
Storage Temperature: Ambient
MDL Number: MFCD00011438
CAS Number: 7719-12-2
EINECS: 231-749-3
UN: 1809
ADR: 6.1 I
Merck Index: 12,07515

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR