Phosphoryl tribromide ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Phosphorus(V) oxybromide, Phosphorus (V) oxide bromide, Phosphoryl bromide, Phosphorus oxybromide
93-1586-100G
93-1586-25G
100203-400EA
1203.08
USD
100203-400
100203-402
Phosphoryl tribromide ≥99%
Phosphoryl tribromide
Formula:
Br₃OP MW: 286.69 g/mol Boiling Pt: 193 °C (1013 hPa) Melting Pt: 52 - 55 °C Density: 2.88 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00036298 CAS Number: 7789-59-5 EINECS: 232-177-7 UN: 1939 ADR: 8,II Merck Index: 12,07505 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...