Tetrabutylphosphonium bromide ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Tetra-n-butylphosphonium bromide

93-1579-100G 93-1579-25G
100200-522EA 86.34 USD
100200-522 100207-788
Tetrabutylphosphonium bromide ≥98%
Tetrabutylphosphonium bromide
Formula: C₁₆H₃₆BrP
MW: 339.34 g/mol
Melting Pt: 100…103 °C
MDL Number: MFCD00011853
CAS Number: 3115-68-2
EINECS: 221-487-8
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR