Trimethyl phosphate ≥97%

Supplier: STREM CHEMICALS INC MS

Synonyms: Trimethylphosphate

93-1561-250G 93-1561-50G
100211-220EA 100.49 USD
100211-220 100197-718
Trimethyl phosphate ≥97%
Trimethyl phosphate

Volatile Precursors for CVD

Formula: C₃H₉O₄P
MW: 140.08 g/mol
Boiling Pt: 197.2 °C (1013 hPa)
Melting Pt: -46 °C
Density: 1.21 g/cm³ (20 °C)
Flash Pt: ≥ 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00008348
CAS Number: 512-56-1
EINECS: 208-144-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR