Phosphorus(III) bromide ≥97%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-1550-100G
93-1550-500G
100204-192EA
107.57
USD
100204-192
100209-412
Phosphorus(III) bromide ≥97%
Phosphorus(III) bromide
Formula:
PBr₃ Boiling Pt: 172.9 °C Melting Pt: –40 °C Density: 2.852 (15°) Storage Temperature: Ambient |
MDL Number:
MFCD00011436 CAS Number: 7789-60-8 EINECS: 232-178-2 UN: 1808 ADR: 8,II Merck Index: 12,07514 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...