Diphosphorus pentoxide ≥98% ACS
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Phosphorus(V) oxide, di-Phosphorus pentoxide, Phosphorus pentoxide, Phosphoric anhydride
93-1548-500G
93-1548-2000G
100199-496EA
227.88
USD
100199-496
100207-458
Diphosphorus pentoxide ≥98% ACS
Diphosphorus pentoxide
Formula:
P₂O₅ MW: 141.94 g/mol Boiling Pt: 300 °C (subl.) Melting Pt: 562 Density: 2.39 Storage Temperature: Ambient |
MDL Number:
MFCD00011440 CAS Number: 1314-56-3 EINECS: 215-236-1 UN: 1807 ADR: 8 II Merck Index: 13,07441 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...