Phosphoryl trichloride 98

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Phosphorus (V) oxide chloride, Phosphorus oxychloride, Phosphoryl chloride, Phosphorus(V) oxychloride

93-1543-250G 93-1543-1000G
100209-024EA 94.83 USD
100209-024 100204-674
Phosphoryl trichloride 98
Phosphoryl trichloride

Halides for CVD & ALD, Volatile Precursors for CVD

Formula: POCl₃
MW: 153.33 g/mol
Boiling Pt: 107
Melting Pt: 1
Density: 1.675
Storage Temperature: Ambient
MDL Number: MFCD00011443
CAS Number: 10025-87-3
EINECS: 233-046-7
UN: 1810
ADR: 6.1 I
Merck Index: 12,07506

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR