Silicon ≥99%, powder

Supplier: STREM CHEMICALS INC MS
Warning

93-1496-500G 93-1496-100G
100207-786EA 186.83 USD
100207-786 100208-510
Silicon ≥99%, powder
Silicon
Formula: Si
MW: 28.09 g/mol
Boiling Pt: 2600 °C
Melting Pt: 1420 °C
Density: 2.42
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
UN: 1346
ADR: 4.1 III
Merck Index: 12,08635

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR