Vinyltrichlorosilane ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Trichloro(vinyl)silane

93-1492-50G 93-1492-250G
100197-710EA 41.05 USD
100197-710 100204-182
Vinyltrichlorosilane ≥97%
Vinyltrichlorosilane
Formula: C₂H₃Cl₃Si
MW: 161.49 g/mol
Boiling Pt: 90
Melting Pt: -95
Density: 1.243 (20°)
Flash Pt: 51 °F
MDL Number: MFCD00000482
CAS Number: 75-94-5
EINECS: 200-917-8
UN: 1305
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR