Diphenylsilane ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

93-1485-5G 93-1485-25G
100198-992EA 45.29 USD
100198-992 100205-356
Diphenylsilane ≥97%
Diphenylsilane
Formula: C₁₂H₁₂Si
MW: 184.31 g/mol
Boiling Pt: 95 °C (1013 hPa)
Density: 0.9962 g/cm³ (20 °C)
Flash Pt: 209 °F
Storage Temperature: Ambient
MDL Number: MFCD00003002
CAS Number: 775-12-2
EINECS: 212-271-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR