Tetramethoxysilane ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Tetramethyl orthosilicate

93-1459-100G 93-1459-25G
100202-530EA 161.35 USD
100202-530 100208-504
Tetramethoxysilane ≥98%
Tetramethoxysilane

Metal Alkoxides, Volatile Precursors for CVD

Formula: C₄H₁₂O₄Si
MW: 152.22 g/mol
Boiling Pt: 121
Melting Pt: -2
Density: 1.032
Flash Pt: 84 °F
Storage Temperature: Ambient
MDL Number: MFCD00008341
CAS Number: 681-84-5
EINECS: 211-656-4
UN: 2606
ADR: 6.1 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR