Tetramethylsilane ≥99.9% for NMR spectroscopy

Supplier: STREM CHEMICALS INC MS
Danger

93-1458-25G 93-1458-100G
100203-388EA 147.2 USD
100203-388 100203-386
Tetramethylsilane ≥99.9% for NMR spectroscopy
Tetramethylsilane

Volatile Precursors for CVD

Formula: C₄H₁₂Si
Boiling Pt: 26.6 °C (1013 hPa)
Melting Pt: -99 °C
Density: 0.642 g/cm³ (20 °C)
Flash Pt: -54 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008274
CAS Number: 75-76-3
EINECS: 200-899-1
UN: 2749
ADR: 3,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR