Disodium metasilicate pentahydrate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Sodium metasilicate pentahydrate

93-1448-2000G 93-1448-500G
100206-942EA 188.25 USD
100206-942 100210-198
Disodium metasilicate pentahydrate ≥99%
Disodium metasilicate pentahydrate
Formula: Na₂SiO₃·5H₂O
MW: 212.14 g/mol
Melting Pt: 40…48 °C
Density: 1.749 g/cm³ (20 °C)
MDL Number: MFCD00149176
CAS Number: 10213-79-3
EINECS: 229-912-9
UN: 3253
ADR: 8,III
Merck Index: 13,08716

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR