Silicon dioxide ≥99.7% -100 mesh
Supplier: STREM CHEMICALS INC MS
93-1435-1000G
93-1435-250G
100198-312EA
193.91
USD
100198-312
100205-338
Silicon dioxide ≥99.7% -100 mesh
Silicon dioxide
Formula:
SiO₂ Boiling Pt: 2230 °C (1013 hPa) Melting Pt: 1500 °C Density: 2.2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 7631-86-9 EINECS: 231-545-4 Merck Index: 14,08493 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...