Trichloro(phenyl)silane ≥97%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Phenyltrichlorosilane
93-1419-100G
93-1419-500G
100198-986EA
46.71
USD
100198-986
100210-190
Trichloro(phenyl)silane ≥97%
Trichloro(phenyl)silane
Formula:
C₆H₅Cl₃Si MW: 211.55 g/mol Boiling Pt: 201 °C (1013 hPa) Melting Pt: -40 °C Density: 1.315 g/cm³ (20 °C) Flash Pt: 196 °F |
MDL Number:
MFCD00000479 CAS Number: 98-13-5 EINECS: 202-640-8 UN: 1804 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...