Diethoxy(dimethyl)silane ≥97%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Dimethyldiethoxysilane
93-1406-50G
93-1406-250G
100206-936EA
36.8
USD
100206-936
100196-500
Diethoxy(dimethyl)silane ≥97%
Diethoxy(dimethyl)silane
Formula:
C₆H₁₆O₂Si MW: 148.28 g/mol Boiling Pt: 114 Melting Pt: -87 Density: 0.840 Flash Pt: 53 °F |
MDL Number:
MFCD00009068 CAS Number: 78-62-6 EINECS: 201-127-6 UN: 2380 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...