(3-Chloropropyl)trimethoxysilane ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 3-Chloropropyltrimethoxysilane, 3-(Trimethoxysilyl)propylchloride, Trimethoxy(3-chloropropyl)silane

93-1404-250G 93-1404-50G
100199-488EA 182.58 USD
100199-488 100205-326
(3-Chloropropyl)trimethoxysilane ≥97%
(3-Chloropropyl)trimethoxysilane
Formula: ClH₂CCH₂CH₂Si(OCH₃)₃
MW: 198.72 g/mol
Boiling Pt: 183 °C
Melting Pt: –50 °C
Density: 1.082 g/cm³ (20 °C)
Flash Pt: 173 °F
Storage Temperature: Ambient
MDL Number: MFCD00000997
CAS Number: 2530-87-2
EINECS: 219-787-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR