Butyltrichlorosilane

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: n-Butyltrichlorosilane, Trichloro(butyl)silane

93-1403-10G 93-1403-50G
100201-906EA 65.11 USD
100201-906 100210-188
Butyltrichlorosilane
Butyltrichlorosilane
Formula: C₄H₉Cl₃Si
MW: 191.56 g/mol
Boiling Pt: 142 °C
Density: 1.16 g/cm³ (20 °C)
Flash Pt: 130 °F
MDL Number: MFCD00013605
CAS Number: 7521-80-4
EINECS: 231-381-3
UN: 1747
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR