Triammonium hexafluoroaluminate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Ammonium hexafluoroaluminate
93-1395-10G
93-1395-50G
100210-186EA
62.28
USD
100210-186
100206-934
Triammonium hexafluoroaluminate ≥98%
tri-Ammonium hexafluoroaluminate
Formula:
H₁₂AlF₆N₃ MW: 195.09 g/mol Density: 1.78 g/cm³ (20 °C) |
MDL Number:
MFCD00042144 CAS Number: 7784-19-2 EINECS: 232-052-7 UN: 3288 ADR: 6.1,III Merck Index: 12,00555 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...