Aluminium bromide ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Aluminum bromide

93-1385-50G 93-1385-250G
100198-982EA 89.17 USD
100198-982 100198-980
Aluminium bromide ≥99%
Aluminium bromide
Formula: AlBr₃
MW: 266.69 g/mol
Boiling Pt: 265
Melting Pt: 97
Density: 2.64
Storage Temperature: Ambient
MDL Number: MFCD00003421
CAS Number: 7727-15-3
EINECS: 231-779-7
UN: 1725
ADR: 8 II
Merck Index: 13,00330

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR