Aluminium nitride, agglomerated powder, high purity

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Aluminum nitride

93-1377-500G 93-1377-100G
100201-902EA 462.83 USD
100201-902 100204-176
Aluminium nitride, agglomerated powder, high purity
Aluminium nitride
Formula: AlN
MW: 40.99 g/mol
Boiling Pt: 2517 °C (1013 hPa)
Melting Pt: 1900…2200 °C
Density: 3.257 to 3.267 g/cm³ (160 °C)
MDL Number: MFCD00003429
CAS Number: 24304-00-5
EINECS: 246-140-8
Merck Index: 12,00366

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR