Triisobutylaluminium ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

93-1358-225G 93-1358-100G
100206-142EA 878.95 USD
100206-142 100210-854
Triisobutylaluminium ≥95%
Triisobutylaluminium

Metal Alkyls, Volatile Precursors for CVD

Formula: C₁₂H₂₇Al
Boiling Pt: 73 °C (5 mmHg)
Melting Pt: 4 °C
Density: 0.7979 g/cm³ (20 °C)
MDL Number: MFCD00008929
CAS Number: 100-99-2
EINECS: 202-906-3
UN: 3394
ADR: 4.2,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR