Diisobutylaluminium hydride 20% in hexanes

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Di-i-butylaluminum hydride, DIBAL-H

93-1355-100G 93-1355-500G
100204-174EA 246.28 USD
100204-174 100205-316
Diisobutylaluminium hydride 20% in hexanes
Diisobutylaluminium hydride
Formula: C₈H₁₉Al
MW: 142.22 g/mol
Density: 0.7 g/cm³ (20 °C)
Flash Pt: –9 °F
Storage Temperature: Ambient
MDL Number: MFCD00008928
CAS Number: 1191-15-7
EINECS: 214-729-9
UN: 3394
ADR: 4.3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR