Aluminium sulfate octadecahydrate ≥98% ACS
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Trade termforalum, Aluminum(III) sulphate octadecahydrate, Aluminum sulphate octadecahydrate
93-1350-2000G
93-1350-500G
100204-172EA
1016.25
USD
100204-172
100200-516
Aluminium sulfate octadecahydrate ≥98% ACS
Aluminium sulphate octadecahydrate
Formula:
Al₂(SO₄)₃·18H₂O MW: 666.43 g/mol Melting Pt: 86.5 °C (dec.) Density: 1.69 g/cm³ (20 °C) |
MDL Number:
MFCD00149136 CAS Number: 7784-31-8 EINECS: 233-135-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...