Aluminium isopropoxide ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-1345-250G
93-1345-1000G
100197-696EA
36.8
USD
100197-696
100211-212
Aluminium isopropoxide ≥98%
Aluminium isopropoxide
Alkoxide, Metal Alkoxides, Volatile Precursors for CVD
Formula:
C₉H₂₁AlO₃ Boiling Pt: 140.5 °C (8 mmHg) Melting Pt: 118.5 °C Density: 1.0346 |
MDL Number:
MFCD00008870 CAS Number: 555-31-7 EINECS: 209-090-8 UN: 3178 ADR: 4.1,II Merck Index: 13,00346 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...