Aluminium oxide ≥99%, powder 1-5 µm

Supplier: STREM CHEMICALS INC MS
Warning

93-1339-1000G 93-1339-250G
100211-210EA 308.55 USD
100211-210 100197-692
Aluminium oxide ≥99%, powder 1-5 µm
Aluminium oxide
Formula: Al₂O₃
Boiling Pt: 2980 °C
Melting Pt: 2045 °C
Density: 3.94 g/cm³ (20 °C)
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR