Aluminium oxide activated basic, γ-phase ≥96%

Supplier: STREM CHEMICALS INC MS
Warning

93-1334-500G 93-1334-2000G
100197-690EA 133.05 USD
100197-690 100211-208
Aluminium oxide activated basic, γ-phase ≥96%
Aluminium oxide activated basic, γ-phase
Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980 °C
Melting Pt: 2045 °C
Density: 3,97
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR