Aluminium oxide activated acidic, γ-phase ≥96%

Supplier: STREM CHEMICALS INC MS
Warning

93-1329-2000G 93-1329-500G
100197-194EA 339.69 USD
100197-194 100206-932
Aluminium oxide activated acidic, γ-phase ≥96%
Aluminium oxide activated acidic, γ-phase

CAS: 1344-28-1
Formula: Al2O3
MW: 101.96 g/mol
EINECS: 215-691-6

Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980 °C
Melting Pt: 2045 °C
Density: 3.97
MDL Number: MFCD00003424
CAS Number: 1344-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR