Aluminium nitrate nonahydrate ≥98% ACS

Supplier: STREM CHEMICALS INC MS
Danger

93-1325-2000G 93-1325-100G 93-1325-500G
100197-688EA 285.91 USD
100197-688 100208-500 100198-304
Aluminium nitrate nonahydrate ≥98% ACS
Aluminium nitrate nonahydrate
Formula: Al(NO₃)₃·9H₂O
Melting Pt: 73.5 °C
Density: 1.72 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149132
CAS Number: 7784-27-2
EINECS: 236-751-8
UN: 1438
ADR: 5.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR