Aluminium trilactate ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: L-lactate aluminum, Lactic acid aluminum salt, (±)-Lactic acid aluminium salt, L-lactate aluminum salt, Aluminum lactate, (±)-2-Hydroxypropanoic acid aluminium salt, Aluminium lactate, DL-Lactic acid aluminium salt

93-1319-100G 93-1319-500G
100197-686EA 69.35 USD
100197-686 100202-438
Aluminium trilactate ≥95%
Aluminium trilactate
Formula: (H₃CCH(OH)COO)₃Al
MW: 294.19
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD20529175
CAS Number: 18917-91-4
EINECS: 242-670-9
Merck Index: 13,00347

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR