Magnesium ≥99%, chips

Supplier: STREM CHEMICALS INC MS
Warning

93-1299-1000G 93-1299-50G 93-1299-250G
100201-890EA 191.08 USD
100201-890 100205-306 100203-370
Magnesium ≥99%, chips
Magnesium
Formula: Mg
Boiling Pt: 1107 °C (1013 hPa)
Melting Pt: 651 °C
Density: 1.74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1,III
Merck Index: 13,05675

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR