Magnesium oxide ≥99.5% (Mg basis) -325 mesh
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Magnesia usta
93-1243-500G
93-1243-100G
100206-926EA
253.35
USD
100206-926
100198-964
Magnesium oxide ≥99.5% (Mg basis) -325 mesh
Magnesium oxide
Formula:
MgO MW: 40.3 g/mol Boiling Pt: 3600 Melting Pt: 2800 Density: 3.58 |
MDL Number:
MFCD00011109 CAS Number: 1309-48-4 EINECS: 215-171-9 Merck Index: 13,05700 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...