Magnesium fluoride ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-1225-2000G 93-1225-500G
100203-834EA 539.26 USD
100203-834 100196-478
Magnesium fluoride ≥99%
Magnesium fluoride
Formula: MgF₂
MW: 62.3 g/mol
Boiling Pt: 2239 °C
Melting Pt: 1248 °C
Density: 3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011108
CAS Number: 7783-40-6
EINECS: 231-995-1
Merck Index: 12,05701

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR