Magnesium chloride, (max. 2% H₂O) ≥97.5%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-1221-1000G
93-1221-250G
100196-476EA
86.34
USD
100196-476
100201-874
Magnesium chloride, (max. 2% H₂O) ≥97.5%
Magnesium chloride
Formula:
MgCl₂ MW: 95.21 g/mol Boiling Pt: 1412 Melting Pt: 715 Density: 2.32 Storage Temperature: Ambient |
MDL Number:
MFCD00011106 CAS Number: 7786-30-3 EINECS: 232-094-6 Merck Index: 13,05684 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...