Magnesium bromide, anhydrous ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

93-1218-10G 93-1218-50G
100197-676EA 58.03 USD
100197-676 100197-678
Magnesium bromide, anhydrous ≥98%
Magnesium bromide
Formula: Br₂Mg
MW: 184.11
Melting Pt: 700
Density: 3.72
MDL Number: MFCD00011105
CAS Number: 7789-48-2
EINECS: 232-170-9
Merck Index: 12,05695

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR