Magnesium acetate tetrahydrate ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: Acetic acid magnesium salt tetrahydrate, Magnesium di(acetate) tetrahydrate

93-1216-250G 93-1216-1000G
100198-962EA 94.83 USD
100198-962 100202-436
Magnesium acetate tetrahydrate ≥99%
Magnesium acetate tetrahydrate
Formula: (H₃CCOO)₂Mg·4H₂O
MW: 214.46 g/mol
Melting Pt: 80
Density: 1.454
Storage Temperature: Ambient
MDL Number: MFCD00149214
CAS Number: 16674-78-5
EINECS: 205-554-9
Merck Index: 13,05676

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR