Disodium metasilicate, anhydrous ≥95%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Sodium metasilicate
93-1196-2000G
93-1196-500G
100210-852EA
215.14
USD
100210-852
100201-870
Disodium metasilicate, anhydrous ≥95%
Disodium metasilicate
Formula:
Na₂O₃Si MW: 122.06 g/mol Melting Pt: 1088 °C Density: 1.749 g/cm³ (20 °C) |
MDL Number:
MFCD00003492 CAS Number: 6834-92-0 EINECS: 229-912-9 UN: 3253 ADR: 8,III Merck Index: 12,08788 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...