Disodium sulphide nonahydrate ≥98% ACS

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Sodium sulphide nonahydrate

93-1183-500G 93-1183-100G
100205-286EA 135.88 USD
100205-286 100205-284
Disodium sulphide nonahydrate ≥98% ACS
Disodium sulphide nonahydrate
Formula: Na₂S·9H₂O
Boiling Pt: 920 °C (1013 hPa)
Melting Pt: 49 °C
Density: 1.427 g/cm³ (16 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00149184
CAS Number: 1313-84-4
EINECS: 215-211-5
UN: 1849
ADR: 8,II
REACH: 01-2119513694-38

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR