Sodium perchlorate monohydrate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

93-1170-500G 93-1170-100G
100208-084EA 179.75 USD
100208-084 100201-864
Sodium perchlorate monohydrate ≥98%
Sodium perchlorate monohydrate
Formula: NaClO₄·H₂O
Melting Pt: 130 °C
Density: 2.02 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149164
CAS Number: 7791-07-3
EINECS: 231-511-9
UN: 1502
ADR: 5.1,II
Merck Index: 12,08798

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR