Disodium oxalate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-1165-500G 93-1165-100G
100196-460EA 227.88 USD
100196-460 100210-164
Disodium oxalate ≥99%
Disodium oxalate
Formula: Na₂C₂O₄
Melting Pt: 250…270 °C
Density: 2.27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012465
CAS Number: 62-76-0
EINECS: 200-550-3
UN: 2811
ADR: 6.1,III
Merck Index: 13,08723

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR